Assay Details
| Gene Symbol: agt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316493.1 | NP_001303422.1 | 1 | 332 | 72 | |
| NM_058018.3 | NP_477366.1 | 1 | 332 | 72 | ||
| GenBank mRNA | AF063906.1 | - | 1 | 332 | 72 | |
| AY089255.1 | - | 1 | 296 | 72 | ||
More Information
Set Membership: |
|

agt
Genomic Map