Assay Details
| Gene Symbol: Sbf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058053.4 | NP_477401.2 | 3 - 4 | 321 | 90 | |
| NM_169430.3 | NP_731633.2 | 3 - 4 | 321 | 90 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbf| Gene Symbol: Sbf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058053.4 | NP_477401.2 | 3 - 4 | 321 | 90 | |
| NM_169430.3 | NP_731633.2 | 3 - 4 | 321 | 90 | ||
Set Membership: |
|