Assay Details
| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058067.3 | NP_477415.1 | 3 - 4 | 614 | 65 | |
| GenBank mRNA | AY118330.1 | - | 3 - 4 | 549 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsc1| Gene Symbol: Tsc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058067.3 | NP_477415.1 | 3 - 4 | 614 | 65 | |
| GenBank mRNA | AY118330.1 | - | 3 - 4 | 549 | 65 | |
Set Membership: |
|