Assay Details
| Gene Symbol: Decay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058114.4 | NP_477462.1 | 1 - 2 | 380 | 79 | |
| GenBank mRNA | AF130469.3 | - | 1 - 2 | 345 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Decay| Gene Symbol: Decay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058114.4 | NP_477462.1 | 1 - 2 | 380 | 79 | |
| GenBank mRNA | AF130469.3 | - | 1 - 2 | 345 | 79 | |
Set Membership: |
|