Assay Details
| Gene Symbol: Decay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058114.4 | NP_477462.1 | 2 - 3 | 710 | 62 | |
| GenBank mRNA | AF130469.3 | - | 2 - 3 | 675 | 62 | |
| AY089242.1 | - | 2 - 3 | 1018 | 62 | ||
More Information
Set Membership: |
|

Decay
Genomic Map