Assay Details
| Gene Symbol: Sce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058161.4 | NP_477509.1 | 1 - 2 | 1381 | 78 | |
| GenBank mRNA | AY058535.1 | - | 1 - 2 | 1355 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sce| Gene Symbol: Sce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058161.4 | NP_477509.1 | 1 - 2 | 1381 | 78 | |
| GenBank mRNA | AY058535.1 | - | 1 - 2 | 1355 | 78 | |
Set Membership: |
|