Assay Details
| Gene Symbol: 5-HT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104214.3 | NP_001097684.2 | 2 - 3 | 1600 | 66 | |
| NM_001170034.2 | NP_001163505.1 | 2 - 3 | 1600 | 66 | ||
| NM_001170035.2 | NP_001163506.2 | 2 - 3 | 1461 | 66 | ||
| NM_079499.4 | NP_524223.2 | 2 - 3 | 1600 | 66 | ||
| NM_169011.3 | NP_730859.1 | 2 - 3 | 1600 | 66 | ||
| GenBank mRNA | BT015194.1 | - | 2 - 3 | 1595 | 66 | |
More Information
Set Membership: |
|

5-HT2A
Genomic Map