Assay Details
| Gene Symbol: kkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079509.3 | NP_524233.1 | 5 - 6 | 2023 | 68 | |
| NM_169052.3 | NP_730928.2 | 5 - 6 | 2023 | 68 | ||
| NM_206430.3 | NP_996152.1 | 5 - 6 | 2023 | 68 | ||
More Information
Set Membership: |
|

kkv
Genomic Map