Assay Details
| Gene Symbol: kkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079509.3 | NP_524233.1 | 6 - 7 | 3951 | 73 | |
| NM_206430.3 | NP_996152.1 | 6 - 7 | 3951 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kkv| Gene Symbol: kkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079509.3 | NP_524233.1 | 6 - 7 | 3951 | 73 | |
| NM_206430.3 | NP_996152.1 | 6 - 7 | 3951 | 73 | ||
Set Membership: |
|