Assay Details
| Gene Symbol: kkv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079509.3 | NP_524233.1 | 8 - 9 | 4331 | 65 | |
| NM_169052.3 | NP_730928.2 | 8 - 9 | 4331 | 65 | ||
| NM_206430.3 | NP_996152.1 | 8 - 9 | 4331 | 65 | ||
More Information
Set Membership: |
|

kkv
Genomic Map