Assay Details
| Gene Symbol: kra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079514.3 | NP_524238.1 | 3 - 4 | 468 | 65 | |
| NM_169074.2 | NP_730958.1 | 3 - 4 | 468 | 65 | ||
| NM_169075.2 | NP_730959.1 | 4 - 5 | 701 | 65 | ||
| NM_169076.2 | NP_730960.1 | 4 - 5 | 760 | 65 | ||
| NM_169077.2 | NP_730961.1 | 3 - 4 | 880 | 65 | ||
| NM_169078.2 | NP_730962.1 | 2 - 3 | 555 | 65 | ||
| NM_169079.2 | NP_730963.1 | 3 - 4 | 630 | 65 | ||
| GenBank mRNA | AF325529.1 | - | 3 - 4 | 409 | 65 | |
| AY058505.1 | - | 3 - 4 | 856 | 65 | ||
More Information
Set Membership: |
|

kra
Genomic Map