Assay Details
| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275399.1 | NP_001262328.1 | 1 - 2 | 286 | 70 | |
| NM_001275403.1 | NP_001262332.1 | 1 - 2 | 286 | 70 | ||
| NM_079526.3 | NP_524250.3 | 1 - 2 | 286 | 70 | ||
| GenBank mRNA | AF217960.1 | - | 1 - 2 | 259 | 70 | |
| AF281145.1 | - | 1 - 2 | 286 | 70 | ||
| BT149892.1 | - | 1 - 2 | 250 | 70 | ||
More Information
Set Membership: |
|

Alh
Genomic Map