Assay Details
| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104227.3 | NP_001097697.2 | 3 - 4 | 2208 | 81 | |
| NM_001170065.2 | NP_001163536.1 | 3 - 4 | 2208 | 81 | ||
| NM_001170066.2 | NP_001163537.1 | 3 - 4 | 2208 | 81 | ||
| NM_001275399.1 | NP_001262328.1 | 7 - 8 | 3358 | 81 | ||
| NM_001275400.1 | NP_001262329.1 | 7 - 8 | 3175 | 81 | ||
| NM_001275401.1 | NP_001262330.1 | 3 - 4 | 2208 | 81 | ||
| NM_001275402.1 | NP_001262331.1 | 3 - 4 | 2208 | 81 | ||
| NM_079526.3 | NP_524250.3 | 7 - 8 | 3331 | 81 | ||
| NM_169170.3 | NP_731131.2 | 3 - 4 | 2208 | 81 | ||
| NM_169171.2 | NP_731132.1 | 7 - 8 | 3148 | 81 | ||
| NM_169172.3 | NP_731133.1 | 3 - 4 | 2208 | 81 | ||
| GenBank mRNA | AF217960.1 | - | 7 - 8 | 3304 | 81 | |
| AF281145.1 | - | 7 - 8 | 3331 | 81 | ||
| AY051905.1 | - | 3 - 4 | 2171 | 81 | ||
| BT149892.1 | - | 7 - 8 | 3295 | 81 | ||
More Information
Set Membership: |
|

Alh
Genomic Map