Assay Details
| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104227.3 | NP_001097697.2 | 5 - 6 | 3076 | 67 | |
| NM_001104228.3 | NP_001097698.1 | 5 - 6 | 4099 | 67 | ||
| NM_001170065.2 | NP_001163536.1 | 5 - 6 | 3076 | 67 | ||
| NM_001170066.2 | NP_001163537.1 | 5 - 6 | 3076 | 67 | ||
| NM_001275399.1 | NP_001262328.1 | 9 - 10 | 4226 | 67 | ||
| NM_001275400.1 | NP_001262329.1 | 9 - 10 | 4043 | 67 | ||
| NM_001275401.1 | NP_001262330.1 | 5 - 6 | 3076 | 67 | ||
| NM_001275402.1 | NP_001262331.1 | 5 - 6 | 3076 | 67 | ||
| NM_001275403.1 | NP_001262332.1 | 9 - 10 | 5222 | 67 | ||
| NM_079526.3 | NP_524250.3 | 9 - 10 | 4199 | 67 | ||
| NM_169170.3 | NP_731131.2 | 5 - 6 | 3076 | 67 | ||
| NM_169171.2 | NP_731132.1 | 9 - 10 | 4016 | 67 | ||
| NM_169172.3 | NP_731133.1 | 5 - 6 | 3076 | 67 | ||
| GenBank mRNA | AF217960.1 | - | 9 - 10 | 4172 | 67 | |
| AF281145.1 | - | 9 - 10 | 4199 | 67 | ||
| AY051905.1 | - | 5 - 6 | 3039 | 67 | ||
| BT149892.1 | - | 9 - 10 | 4163 | 67 | ||
More Information
Set Membership: |
|

Alh
Genomic Map