Assay Details
| Gene Symbol: lap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144547.3 | NP_001138019.2 | 4 - 5 | 807 | 63 | |
| NM_001170067.2 | NP_001163538.1 | 4 - 5 | 807 | 63 | ||
| NM_001260029.2 | NP_001246958.1 | 4 - 5 | 807 | 63 | ||
| NM_001260030.2 | NP_001246959.1 | 4 - 5 | 807 | 63 | ||
| NM_001260031.2 | NP_001246960.1 | 4 - 5 | 807 | 63 | ||
| NM_001275410.1 | NP_001262339.1 | 4 - 5 | 807 | 63 | ||
| NM_079528.3 | NP_524252.2 | 4 - 5 | 807 | 63 | ||
More Information
Set Membership: |
|

lap
Genomic Map