Assay Details
| Gene Symbol: dsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275423.1 | NP_001262352.1 | 2 - 3 | 1928 | 113 | |
| NM_001275424.1 | NP_001262353.1 | 2 - 3 | 2317 | 113 | ||
| NM_001300291.1 | NP_001287220.1 | 2 - 3 | 2317 | 113 | ||
| NM_079548.5 | NP_524272.4 | 2 - 3 | 1928 | 113 | ||
| NM_169202.1 | NP_731197.1 | 2 - 3 | 2317 | 113 | ||
| NM_169203.2 | NP_731198.1 | 2 - 3 | 2317 | 113 | ||
| GenBank mRNA | BT029258.1 | - | 2 - 3 | 1928 | 113 | |
More Information
Set Membership: |
|

dsx
Genomic Map