Assay Details
| Gene Symbol: puc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300294.1 | NP_001287223.1 | 3 - 4 | 627 | 59 | |
| NM_079549.3 | NP_524273.1 | 3 - 4 | 1139 | 59 | ||
| GenBank mRNA | AY061616.1 | - | 3 - 4 | 1083 | 59 | |
More Information
Set Membership: |
|

puc
Genomic Map