Assay Details
| Gene Symbol: RpA-70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079550.4 | NP_524274.1 | 2 - 3 | 369 | 71 | |
| GenBank mRNA | AY069331.1 | - | 2 - 3 | 323 | 71 | |
| Z70277.1 | - | 2 - 3 | 294 | 71 | ||
More Information
Set Membership: |
|

RpA-70
Genomic Map