Assay Details
| Gene Symbol: stck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260058.2 | NP_001246987.1 | 5 - 6 | 803 | 66 | |
| NM_079554.4 | NP_524278.1 | 4 - 5 | 683 | 66 | ||
| NM_169220.3 | NP_731241.1 | 4 - 5 | 691 | 66 | ||
| NM_169221.3 | NP_731242.1 | 4 - 5 | 642 | 66 | ||
| GenBank mRNA | AF078907.1 | - | 4 - 5 | 650 | 66 | |
| AY060424.1 | - | 3 - 4 | 2002 | 66 | ||
| BT003779.1 | - | 4 - 5 | 694 | 66 | ||
| BT012325.1 | - | 4 - 5 | 634 | 66 | ||
| BT044500.1 | - | 4 - 5 | 689 | 66 | ||
More Information
Set Membership: |
|

stck
Genomic Map