Assay Details
| Gene Symbol: stck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260058.2 | NP_001246987.1 | 6 - 7 | 1065 | 63 | |
| NM_079554.4 | NP_524278.1 | 5 - 6 | 945 | 63 | ||
| NM_169220.3 | NP_731241.1 | 5 - 6 | 953 | 63 | ||
| NM_169221.3 | NP_731242.1 | 5 - 6 | 904 | 63 | ||
| GenBank mRNA | AF078907.1 | - | 5 - 6 | 912 | 63 | |
| AY060424.1 | - | 4 - 5 | 2264 | 63 | ||
| BT003779.1 | - | 5 - 6 | 956 | 63 | ||
| BT012325.1 | - | 5 - 6 | 896 | 63 | ||
| BT044500.1 | - | 5 - 6 | 951 | 63 | ||
More Information
Set Membership: |
|

stck
Genomic Map