Assay Details
| Gene Symbol: Calr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275501.1 | NP_001262430.1 | 3 - 4 | 750 | 67 | |
| NM_079569.5 | NP_524293.2 | 3 - 4 | 750 | 67 | ||
| GenBank mRNA | BT001670.1 | - | 3 - 4 | 742 | 67 | |
More Information
Set Membership: |
|

Calr
Genomic Map