Assay Details
| Gene Symbol: pros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260117.2 | NP_001247046.1 | 5 - 6 | 6251 | 106 | |
| NM_079593.6 | NP_524317.4 | 2 - 3 | 4006 | 106 | ||
| GenBank mRNA | Z11743.1 | - | 4004 | 106 | ||
More Information
Set Membership: |
|

pros
Genomic Map