Assay Details
| Gene Symbol: pros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260115.2 | NP_001247044.1 | 6 - 7 | 6313 | 78 | |
| NM_001260116.2 | NP_001247045.1 | 5 - 6 | 7630 | 78 | ||
| NM_001260117.2 | NP_001247046.1 | 6 - 7 | 6433 | 78 | ||
| NM_079593.6 | NP_524317.4 | 3 - 4 | 4188 | 78 | ||
| NM_169386.3 | NP_731565.4 | 2 - 3 | 4584 | 78 | ||
| NM_176459.4 | NP_788636.3 | 3 - 4 | 4101 | 78 | ||
| GenBank mRNA | BT015263.1 | - | 3 - 4 | 4087 | 78 | |
| Z11743.1 | - | 4186 | 78 | |||
More Information
Set Membership: |
|

pros
Genomic Map