Assay Details
| Gene Symbol: pros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260115.2 | NP_001247044.1 | 7 - 8 | 6456 | 73 | |
| NM_001260116.2 | NP_001247045.1 | 6 - 7 | 7773 | 73 | ||
| NM_001260117.2 | NP_001247046.1 | 7 - 8 | 6576 | 73 | ||
| NM_079593.6 | NP_524317.4 | 4 - 5 | 4331 | 73 | ||
| NM_169386.3 | NP_731565.4 | 3 - 4 | 4727 | 73 | ||
| NM_176459.4 | NP_788636.3 | 4 - 5 | 4244 | 73 | ||
| GenBank mRNA | BT015263.1 | - | 4 - 5 | 4230 | 73 | |
| Z11743.1 | - | 4329 | 73 | |||
More Information
Set Membership: |
|

pros
Genomic Map