Assay Details
| Gene Symbol: Fer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079598.3 | NP_524322.1 | 1 - 2 | 230 | 101 | |
| GenBank mRNA | AF517123.1 | - | 1 - 2 | 200 | 101 | |
| AY094777.1 | - | 1 - 2 | 196 | 101 | ||
More Information
Set Membership: |
|

Fer3
Genomic Map