Assay Details
| Gene Symbol: Prosalpha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079604.3 | NP_524328.1 | 3 - 4 | 495 | 59 | |
| GenBank mRNA | AY069280.1 | - | 3 - 4 | 433 | 59 | |
More Information
Set Membership: |
|

Prosalpha2
Genomic Map