Assay Details
| Gene Symbol: DNApol-theta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079609.3 | NP_524333.1 | 1 - 2 | 1678 | 69 | |
| GenBank mRNA | BT044169.1 | - | 1 - 2 | 1678 | 69 | |
More Information
Set Membership: |
|

DNApol-theta
Genomic Map