Assay Details
| Gene Symbol: CtBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014617.2 | NP_001014617.1 | 4 - 5 | 1145 | 79 | |
| NM_001275591.2 | NP_001262520.1 | 4 - 5 | 1145 | 79 | ||
| NM_001275592.1 | NP_001262521.1 | 4 - 5 | 1132 | 79 | ||
| NM_001275593.1 | NP_001262522.1 | 4 - 5 | 1145 | 79 | ||
| NM_001275594.1 | NP_001262523.1 | 4 - 5 | 1145 | 79 | ||
| NM_001275595.1 | NP_001262524.1 | 4 - 5 | 1145 | 79 | ||
| NM_079612.3 | NP_524336.2 | 4 - 5 | 1145 | 79 | ||
| NM_169490.2 | NP_731762.1 | 4 - 5 | 1285 | 79 | ||
| NM_169491.2 | NP_731763.1 | 4 - 5 | 1569 | 79 | ||
| NM_169492.2 | NP_731764.1 | 4 - 5 | 1132 | 79 | ||
| GenBank mRNA | AY069170.1 | - | 4 - 5 | 1285 | 79 | |
More Information
Set Membership: |
|

CtBP
Genomic Map