Assay Details
| Gene Symbol: sim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079616.4 | NP_524340.2 | 1 - 2 | 151 | 68 | |
| GenBank mRNA | AY129457.1 | - | 1 - 2 | 148 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sim| Gene Symbol: sim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079616.4 | NP_524340.2 | 1 - 2 | 151 | 68 | |
| GenBank mRNA | AY129457.1 | - | 1 - 2 | 148 | 68 | |
Set Membership: |
|