Assay Details
| Gene Symbol: timeout | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079617.3 | NP_524341.3 | 2 - 3 | 541 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

timeout| Gene Symbol: timeout | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079617.3 | NP_524341.3 | 2 - 3 | 541 | 86 | |
Set Membership: |
|