Assay Details
| Gene Symbol: Su(var)3-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079633.3 | NP_524357.2 | 2 - 3 | 357 | 74 | |
| GenBank mRNA | BT133458.1 | - | 2 - 3 | 337 | 74 | |
More Information
Set Membership: |
|

Su(var)3-9
Genomic Map