Assay Details
| Gene Symbol: Tm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275663.1 | NP_001262592.1 | 2 - 3 | 733 | 76 | |
| NM_001275664.1 | NP_001262593.1 | 2 - 3 | 1051 | 76 | ||
| NM_001300408.1 | NP_001287337.1 | 1 - 2 | 678 | 76 | ||
| NM_079637.5 | NP_524361.4 | 2 - 3 | 761 | 76 | ||
| NM_169644.2 | NP_732012.1 | 2 - 3 | 814 | 76 | ||
| NM_169645.4 | NP_732013.2 | 2 - 3 | 709 | 76 | ||
| GenBank mRNA | BT044580.1 | - | 2 - 3 | 761 | 76 | |
| BT133082.1 | - | 1 - 2 | 668 | 76 | ||
More Information
Set Membership: |
|

Tm2
Genomic Map