Assay Details
| Gene Symbol: glob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300414.2 | NP_001287343.1 | 2 - 3 | 383 | 71 | |
| NM_079645.4 | NP_524369.1 | 2 - 3 | 416 | 71 | ||
| NM_169678.2 | NP_732081.1 | 2 - 3 | 614 | 71 | ||
| NM_169679.3 | NP_732082.1 | 2 - 3 | 383 | 71 | ||
| NM_169680.3 | NP_732083.1 | 3 - 4 | 565 | 71 | ||
| GenBank mRNA | BT001819.1 | - | 2 - 3 | 298 | 71 | |
| KX531616.1 | - | 1 - 2 | 159 | 71 | ||
More Information
Set Membership: |
|

glob1
Genomic Map