Assay Details
| Gene Symbol: Fer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300429.1 | NP_001287358.1 | 1 - 2 | 137 | 65 | |
| NM_001300430.1 | NP_001287359.1 | 1 - 2 | 137 | 65 | ||
| NM_001300431.1 | NP_001287360.1 | 1 - 2 | 137 | 65 | ||
| NM_079652.3 | NP_524376.2 | 1 - 2 | 137 | 65 | ||
More Information
Set Membership: |
|

Fer2
Genomic Map