Assay Details
| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079673.3 | NP_524397.2 | 7 - 8 | 3930 | 135 | |
| NM_169816.2 | NP_732344.1 | 7 - 8 | 2329 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fru| Gene Symbol: fru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079673.3 | NP_524397.2 | 7 - 8 | 3930 | 135 | |
| NM_169816.2 | NP_732344.1 | 7 - 8 | 2329 | 135 | ||
Set Membership: |
|