Assay Details
| Gene Symbol: cdi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260262.2 | NP_001247191.1 | 6 - 7 | 2136 | 110 | |
| NM_079677.4 | NP_524401.2 | 6 - 7 | 2139 | 110 | ||
| GenBank mRNA | BT004492.1 | - | 5 - 6 | 1185 | 110 | |
More Information
Set Membership: |
|

cdi
Genomic Map