Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043257.2 | NP_001036722.1 | 13 - 14 | 6975 | 65 | |
| NM_001043258.2 | NP_001036723.1 | 13 - 14 | 6975 | 65 | ||
| NM_001043259.2 | NP_001036724.1 | 12 - 13 | 5976 | 65 | ||
| NM_001043261.2 | NP_001036726.1 | 12 - 13 | 6688 | 65 | ||
| NM_001043263.2 | NP_001036728.1 | 12 - 13 | 6902 | 65 | ||
| NM_001275801.1 | NP_001262730.1 | 11 - 12 | 5861 | 65 | ||
| NM_001275803.1 | NP_001262732.1 | 12 - 13 | 5976 | 65 | ||
| NM_001275804.1 | NP_001262733.1 | 2 - 3 | 519 | 65 | ||
| GenBank mRNA | AF277386.1 | - | 11 - 12 | 5545 | 65 | |
More Information
Set Membership: |
|

Dys
Genomic Map