Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 6 - 7 | 2120 | 75 | |
| NM_001043257.2 | NP_001036722.1 | 22 - 23 | 7967 | 75 | ||
| NM_001043258.2 | NP_001036723.1 | 22 - 23 | 7967 | 75 | ||
| NM_001043259.2 | NP_001036724.1 | 20 - 21 | 6899 | 75 | ||
| NM_001043261.2 | NP_001036726.1 | 21 - 22 | 7680 | 75 | ||
| NM_001043263.2 | NP_001036728.1 | 21 - 22 | 7894 | 75 | ||
| NM_001275801.1 | NP_001262730.1 | 19 - 20 | 6784 | 75 | ||
| NM_001275803.1 | NP_001262732.1 | 20 - 21 | 6899 | 75 | ||
| NM_001275804.1 | NP_001262733.1 | 10 - 11 | 1442 | 75 | ||
| GenBank mRNA | AF277386.1 | - | 19 - 20 | 6468 | 75 | |
More Information
Set Membership: |
|

Dys
Genomic Map