Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 9 - 10 | 2573 | 61 | |
| NM_001043257.2 | NP_001036722.1 | 25 - 26 | 8420 | 61 | ||
| NM_001043258.2 | NP_001036723.1 | 25 - 26 | 8420 | 61 | ||
| NM_001043259.2 | NP_001036724.1 | 23 - 24 | 7352 | 61 | ||
| NM_001043260.2 | NP_001036725.1 | 5 - 6 | 3356 | 61 | ||
| NM_001043261.2 | NP_001036726.1 | 24 - 25 | 8133 | 61 | ||
| NM_001043263.2 | NP_001036728.1 | 24 - 25 | 8347 | 61 | ||
| NM_001275801.1 | NP_001262730.1 | 22 - 23 | 7237 | 61 | ||
| NM_001275803.1 | NP_001262732.1 | 23 - 24 | 7352 | 61 | ||
| NM_001275804.1 | NP_001262733.1 | 13 - 14 | 1895 | 61 | ||
| GenBank mRNA | AF277386.1 | - | 22 - 23 | 6921 | 61 | |
More Information
Set Membership: |
|

Dys
Genomic Map