Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 11 - 12 | 2993 | 66 | |
| NM_001043257.2 | NP_001036722.1 | 27 - 28 | 8840 | 66 | ||
| NM_001043258.2 | NP_001036723.1 | 27 - 28 | 8840 | 66 | ||
| NM_001043259.2 | NP_001036724.1 | 25 - 26 | 7772 | 66 | ||
| NM_001043260.2 | NP_001036725.1 | 7 - 8 | 3776 | 66 | ||
| NM_001043261.2 | NP_001036726.1 | 26 - 27 | 8553 | 66 | ||
| NM_001043262.2 | NP_001036727.1 | 3 - 4 | 2000 | 66 | ||
| NM_001043263.2 | NP_001036728.1 | 26 - 27 | 8767 | 66 | ||
| NM_001275801.1 | NP_001262730.1 | 25 - 26 | 7708 | 66 | ||
| NM_001275802.1 | NP_001262731.1 | 3 - 4 | 1599 | 66 | ||
| NM_001275803.1 | NP_001262732.1 | 26 - 27 | 7823 | 66 | ||
| NM_001275804.1 | NP_001262733.1 | 16 - 17 | 2366 | 66 | ||
| GenBank mRNA | AF277386.1 | - | 24 - 25 | 7341 | 66 | |
| BT010020.1 | - | 3 - 4 | 1705 | 66 | ||
More Information
Set Membership: |
|

Dys
Genomic Map