Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 12 - 13 | 3193 | 84 | |
| NM_001043257.2 | NP_001036722.1 | 28 - 29 | 9040 | 84 | ||
| NM_001043258.2 | NP_001036723.1 | 28 - 29 | 9040 | 84 | ||
| NM_001043259.2 | NP_001036724.1 | 26 - 27 | 7972 | 84 | ||
| NM_001043260.2 | NP_001036725.1 | 8 - 9 | 3976 | 84 | ||
| NM_001043261.2 | NP_001036726.1 | 27 - 28 | 8753 | 84 | ||
| NM_001043262.2 | NP_001036727.1 | 4 - 5 | 2200 | 84 | ||
| NM_001043263.2 | NP_001036728.1 | 27 - 28 | 8967 | 84 | ||
| NM_001275801.1 | NP_001262730.1 | 26 - 27 | 7908 | 84 | ||
| NM_001275802.1 | NP_001262731.1 | 4 - 5 | 1799 | 84 | ||
| NM_001275803.1 | NP_001262732.1 | 27 - 28 | 8023 | 84 | ||
| NM_001275804.1 | NP_001262733.1 | 17 - 18 | 2566 | 84 | ||
| GenBank mRNA | AF277386.1 | - | 25 - 26 | 7541 | 84 | |
| BT010020.1 | - | 4 - 5 | 1905 | 84 | ||
More Information
Set Membership: |
|

Dys
Genomic Map