Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 13 - 14 | 3328 | 63 | |
| NM_001043257.2 | NP_001036722.1 | 29 - 30 | 9175 | 63 | ||
| NM_001043258.2 | NP_001036723.1 | 29 - 30 | 9175 | 63 | ||
| NM_001043259.2 | NP_001036724.1 | 27 - 28 | 8107 | 63 | ||
| NM_001043260.2 | NP_001036725.1 | 9 - 10 | 4111 | 63 | ||
| NM_001043261.2 | NP_001036726.1 | 28 - 29 | 8888 | 63 | ||
| NM_001043262.2 | NP_001036727.1 | 5 - 6 | 2335 | 63 | ||
| NM_001043263.2 | NP_001036728.1 | 28 - 29 | 9102 | 63 | ||
| NM_001275801.1 | NP_001262730.1 | 27 - 28 | 8043 | 63 | ||
| NM_001275802.1 | NP_001262731.1 | 5 - 6 | 1934 | 63 | ||
| NM_001275803.1 | NP_001262732.1 | 28 - 29 | 8158 | 63 | ||
| NM_001275804.1 | NP_001262733.1 | 18 - 19 | 2701 | 63 | ||
| GenBank mRNA | AF277386.1 | - | 26 - 27 | 7676 | 63 | |
| BT010020.1 | - | 5 - 6 | 2040 | 63 | ||
More Information
Set Membership: |
|

Dys
Genomic Map