Assay Details
| Gene Symbol: Dys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043256.2 | NP_001036721.1 | 18 - 19 | 4915 | 69 | |
| NM_001043258.2 | NP_001036723.1 | 34 - 35 | 10762 | 69 | ||
| NM_001043259.2 | NP_001036724.1 | 32 - 33 | 9694 | 69 | ||
| NM_001043260.2 | NP_001036725.1 | 14 - 15 | 5698 | 69 | ||
| NM_001043261.2 | NP_001036726.1 | 33 - 34 | 10475 | 69 | ||
| NM_001043262.2 | NP_001036727.1 | 10 - 11 | 3922 | 69 | ||
| NM_001043263.2 | NP_001036728.1 | 33 - 34 | 10689 | 69 | ||
| NM_001275803.1 | NP_001262732.1 | 33 - 34 | 9745 | 69 | ||
| NM_001275804.1 | NP_001262733.1 | 23 - 24 | 4288 | 69 | ||
| GenBank mRNA | AF277386.1 | - | 31 - 32 | 9263 | 69 | |
| BT010020.1 | - | 10 - 11 | 3627 | 69 | ||
More Information
Set Membership: |
|

Dys
Genomic Map