Assay Details
| Gene Symbol: ort | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079682.3 | NP_524406.1 | 2 - 3 | 421 | 62 | |
| GenBank mRNA | AF382403.1 | - | 2 - 3 | 402 | 62 | |
| AF411340.1 | - | 2 - 3 | 421 | 62 | ||
| AF435469.1 | - | 1 - 2 | 90 | 62 | ||
| AY049774.1 | - | 2 - 3 | 421 | 62 | ||
| BT029033.1 | - | 2 - 3 | 397 | 62 | ||
More Information
Set Membership: |
|

ort
Genomic Map