Assay Details
| Gene Symbol: Calx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260283.2 | NP_001247212.1 | 10 - 11 | 3259 | 56 | |
| NM_001260284.2 | NP_001247213.1 | 9 - 10 | 3114 | 56 | ||
| NM_001260285.2 | NP_001247214.1 | 9 - 10 | 3066 | 56 | ||
| NM_001260286.2 | NP_001247215.1 | 10 - 11 | 3977 | 56 | ||
| NM_001300505.1 | NP_001287434.1 | 9 - 10 | 3219 | 56 | ||
| NM_001300506.1 | NP_001287435.1 | 9 - 10 | 4633 | 56 | ||
| NM_079699.4 | NP_524423.2 | 8 - 9 | 3908 | 56 | ||
| NM_169940.4 | NP_732576.1 | 9 - 10 | 3311 | 56 | ||
| NM_169941.2 | NP_732577.1 | 8 - 9 | 3153 | 56 | ||
| GenBank mRNA | AY118680.1 | - | 9 - 10 | 3257 | 56 | |
| BT011420.1 | - | 9 - 10 | 3254 | 56 | ||
More Information
Set Membership: |
|

Calx
Genomic Map