Assay Details
| Gene Symbol: Dhc93AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275864.1 | NP_001262793.1 | 22 - 23 | 11211 | 101 | |
| NM_079700.3 | NP_524424.2 | 21 - 22 | 11181 | 101 | ||
More Information
Set Membership: |
|

Dhc93AB
Genomic Map