Assay Details
Gene Symbol: Dhc93AB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275864.1 | NP_001262793.1 | 25 - 26 | 12123 | 64 | |
NM_079700.3 | NP_524424.2 | 24 - 25 | 12093 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
