Assay Details
| Gene Symbol: Dhc93AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275864.1 | NP_001262793.1 | 8 - 9 | 3991 | 71 | |
| NM_079700.3 | NP_524424.2 | 8 - 9 | 3991 | 71 | ||
More Information
Set Membership: |
|

Dhc93AB
Genomic Map