Assay Details
| Gene Symbol: InR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144621.3 | NP_001138093.1 | 4 - 5 | 2131 | 67 | |
| NM_001144622.2 | NP_001138094.1 | 3 - 4 | 1432 | 67 | ||
| NM_001144623.2 | NP_001138095.1 | 3 - 4 | 1498 | 67 | ||
| NM_079712.6 | NP_524436.2 | 4 - 5 | 2141 | 67 | ||
More Information
Set Membership: |
|

InR
Genomic Map