Assay Details
| Gene Symbol: InR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144621.3 | NP_001138093.1 | 11 - 12 | 4723 | 94 | |
| NM_001144622.2 | NP_001138094.1 | 10 - 11 | 4024 | 94 | ||
| NM_001144623.2 | NP_001138095.1 | 10 - 11 | 4090 | 94 | ||
| NM_079712.6 | NP_524436.2 | 11 - 12 | 4733 | 94 | ||
More Information
Set Membership: |
|

InR
Genomic Map